Would you like to log in again?
Multiwipes™ 1-Ply Tissues
Supplier: TECHSOURCE BE
Multiwipes™
8091 8091
490010-900BX 210.12 USD InStock
490010-900 490010-901
Multiwipes™ 1-Ply Tissues
Wipes Disposable Wipes and Towels Wipes for Delicate Tasks
These low lint 1-ply multiwipes made from virgin cellulose are designed for cleaning critical surfaces, and blotting liquids from liquid handling equipment such as pipettors.
Although multiwipes are used specifically for lab and technical applications, they are competitively priced.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty