Would you like to log in again?
Color Coded Tough-Tags, Diversified Biotech
Supplier: Diversified Biotech
COLR-1000 COLB-1000 COLG-1000
490006-710EA 50.99 USD InStock
490006-710 490006-706 490006-708
Color Coded Tough-Tags, Diversified Biotech
Labels
Color Coded Tough-Tagswill not come off in rotors, cyclers or racks. They are resistant to extreme temperatures and the matte surface is suitable for pencil, pen or marker.
  • With 0.44" spot (11 mm)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty