Would you like to log in again?
Cryogenic Storage Dots and Strips,
Supplier: Nev's Ink
L-5001-B L-5000-9 L-5002-1Y
490002-832CS 90.76 USD InStock
490002-832 490002-868 490002-816
Cryogenic Storage Dots and Strips,
Labels
GeneMate cryogenic storage dots and strips are full line of colors and sizes which is perfect for color coding and distinguishing various specimens.
  • Ideal for extreme conditions
  • Advanced adhesive withstands extreme cold, to –196 °C (–311 °F)
  • Suitable for laser printing or hand-writing - will not bleed
  • Available in rolls or laser sheets
Delivery information: Laser labels include template for printing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty