Would you like to log in again?
Advansta WesternBright ChemiPen
Supplier: Advansta
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
R-07055-001
490005-054EA 183.11 USD InStock
490005-054
Advansta WesternBright ChemiPen
Pens Lab Markers
  • Improve accuracy
  • Transform your visible protein ladder standard into a chemiluminescent standard on your Western blot
  • AnnotatePermanently mark your blots with the date or blot ID
  • Confirm HRP substrate stabilityThe ChemiPen reagent glows when incubated with an HRP substrate, so it can be used to test substrate stability
  • Compatible with both x-ray film and CCD imagers
Transform your protein standard into a chemiluminescent marker. Write or draw on your transfer membranes with the WesternBright ChemiPen. With the proprietary "ink" you can make pre-stained protein standards chemiluminescent, annotate your blot with a date or blot number, or check the quality of your HRP substrates. The reagent in the ChemiPen adsorbs to nitrocellulose and PVDF membranes, and reacts with HRP substrates to produce chemiluminescence that can be detected with X-ray.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty