Would you like to log in again?
SP Bel-Art Octagon Spinbar Magnetic Stirring Bars, Bel-Art Products, a part of SP
Supplier: Bel-Art
Spinbar®
F37110-0178 F37110-1122 F37110-0078 F37110-1256 F37110-5838 F37110-0118 F37110-0003 F37110-0058 F37110-7838 F37110-0138 F37110-1128 F37110-0002 F37110-1388 F37110-0112 F37110-0158 F37110-0212 F37110-1218 F37110-0238 F37110-0001 F37110-1238 F37110-2128
490005-622EA 11.86 USD InStock
490005-622 490005-600 490005-610 490005-598 490005-620 54809-770 490005-626 490005-604 490005-614 490005-602 490005-624 490005-612 490005-608 490005-618 490005-628 490005-606 490005-616 490005-592 490005-590 490005-596 490005-594
SP Bel-Art Octagon Spinbar Magnetic Stirring Bars, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
Octagon magnetic stirring bars with integral pivot ring are the most commonly used shape.
  • PTFE low-friction coating
  • Durable and inert
  • Interrupted profile provides greater surface area and added turbulence
  • Pivot ring aids in reducing friction and chattering (F37110-1218 does not have a pivot ring)
  • Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty