Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GeneMate Mini Dry Bath

Supplier: Benchmark Scientific
BSH200-GM
490000-068EA 466.89 USD InStock
490000-068
GeneMate Mini Dry Bath
Block Heaters Digital Block Heaters
The Mini Dry Bath is a personal sized instument that fits in the palm of your hand. Six block configurations are available for a variety of tube sizes, ranging from 0.2 to 50 ml.

  • 2-year full replacement warranty
  • Compact, fits in the palm of your hand (less than 4.4" wide)
  • Exchangeable blocks, for tubes 0.2 to 50 ml
  • Clear cover ensures temperature uniformity
  • Simple touch pad control with digital display
  • Digital temperature control
  • Portable (optional 12 V car adapter)
  • Blocks Sold Separately

A large, backlit display is used to control temperature up to 100 °C (minimum temperature is ambient 5 °C). Each unit includes a clear plastic cover that provides enhanced temperature uniformity and ramping time. A convenient block lifter is also included, allowing block to be removed from the instrument before they have cooled.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty