Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

GeneMate Orbital Shaker, Benchmark Scientific

Supplier: Benchmark Scientific
GeneMate
GM3000
490000-128EA 4078.32 USD InStock
490000-128
GeneMate Orbital Shaker, Benchmark Scientific
Shakers and Mixers Orbital Shakers
The powerful Orbital Shaker is capable of mixing a 14 lb. load capacity up to speeds as high as 300 rpm. The standard platform (included) features a non-slip, rubber coated surface, ideal for tissue culture flasks, Petri dishes and staining trays.

  • 2 year full replacement warranty
  • Large platform, up to 24 x 125 mL, 14 x 250 mL, 9 x 500 mL, 5 x 1 L, or 2 x 2 L
  • Digital speed control up to 300 rpm
  • Equipped with 13" x 12" non-skid rubber coated platform
  • Standard platform dimensions: 33 x 30 cm (13 x 12")
  • MAGic Clamp Platform: 35 x 30 cm (14 x 12")

The convenient MAGic Clamp platform (patent pending) is also available for use with Erlenmeyer flasks and tube racks. The unique, magnetic attachment method is the easiest way to instantly change between flask clamps of different sizes.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty