Would you like to log in again?
Biochrom Colourwave CO7500 Educational Colorimeter
Supplier: Biochrom
80-3000-43 80-3000-44
490005-902EA 2148.08 USD InStock
490005-902 490005-904
Biochrom Colourwave CO7500 Educational Colorimeter
Colorimeters Multiparameter Colorimeters
Biochrom has helped to educate scientists in spectrophotometry for over 30 years. Our Biochrom WPA brand of spectrophotometers and colorimeters are used by schools, colleges and universities worldwide to educate the next generation of scientists.
  • Single beam filter based colorimeter covering 440 - 680 nm
  • Rugged, portable, easy to use
  • Excellent value for money, ideal for educational applications
  • Absorbance, %Transmission and simple kinetic measurements
The Biochrom WPA CO7500 colorimeter is a value for money instrument that has been designed for use in educational establishments such as schools and colleges. It is compact, lightweight and ergonomically designed to withstand student use! This makes it ideal for use in busy teaching laboratories. The large display is easy to read and the instrument is simple to both set up and use, giving rapid results. The samples can be measured in either standard or semi micro 10mm pathlength cuvettes or in 16mm diameter test tubes (adapters for 10/12 mm test tubes are available). Available in mains only or mains /internal rechargeable NiMH battery versions.
Measures Absorbance, % Transmission and simple kinetics. When used in kinetics mode - to study rates of reaction, for example - the CO7500 colorimeter takes readings approximately every second and these can be sent to a chart recorder (via the analogue output) if required. Alternatively, the results could be downloaded to an Excel spreadsheet for graphing and other manipulation using the appropriate accessories.
The CO7500 colorimeter is designed to make life easier for technical staff, and to be low maintainance under demanding conditions. The eight filters (440, 470, 490, 520, 550, 580, 600 and 680nm) are encased in an integral filter wheel so filters cannot be accidentally lost or damaged. With no lid needed, this robust instrument is easy to use, clean and store. There is a drain hole at the bottom of the cell compartment so that spillages do not affect the instrument. Lamp life is extended as it is only on during the measurement cycle. Spare lamps are available and easy to install.
Ordering information: Rechargeable battery version available (490005-904).
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty