Would you like to log in again?
Aluminum Sealing Foils- for PCR and Cold Storage, GeneMate
Supplier: GeneMate
F-GM-100
490001-216PK 158.69 USD InStock
490001-216
Aluminum Sealing Foils- for PCR and Cold Storage, GeneMate
Seals Microplate Seals
These 38 m thick dead-soft aluminum foils with proprietary chemically-resistant inert acrylic adhesive are ideal for PCR as well as storage and sealing of light sensitive compounds or reagents.
Each foil has a single end tab, removable at perforation. GeneMate Aluminum Sealing Foil eliminates the need for heat-sealing devices that can affect the integrity of the sample or distort the plate during thermal cycling. Foil sheets are also a more user friendly alternative to sealing mats. Each foil sheet measures 126.72 x 78.23 mm and offers sufficient sealing area for all PCR plates. The full length perforated end tab allows for easy alignment and application to the plate plus easy tab removal. Length between the perforations with end tabs removed is 116.21 mm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty