Would you like to log in again?
Oakton® Acorn Meters, Cole Parmer
Supplier: Cole Parmer
WD-35613-52 WD-35613-22
101076-424EA 499.45 USD InStock
101076-424 101076-428
Oakton® Acorn Meters, Cole Parmer
Electrochemical Meters pH and Multi-Parameter Meters
pH range: -1.0 to 15.0, resolution: 0.1 pH, accuracy: ±0.1 pH.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty