Would you like to log in again?
HyperREZ™ XP Carboyhydrate Na⁺ HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
69310-057726 69310-307780
10046-260EA 3021.85 USD InStock
10046-260 10046-258
HyperREZ™ XP Carboyhydrate Na⁺ HPLC Columns, Thermo Scientific
Chromatography Columns
HyperREZ XP Carbohydrate columns are based on a monodisperse resin with a 4 or 8% divinylbenzene content, and provide an ideal medium for the analysis of carbohydrates and organic acids.
Unlike silica based columns they are stable at low pH, allowing the use of dilute acid as a mobile phase. The columns can also be run at elevated temperatures, for faster analysis and improved resolution of some closely eluting analytes.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty