Would you like to log in again?
EarlyTox Cardiotoxicity Kit, Molecular Devices
Supplier: Molecular Devices
R8211 R8211
10048-824EA 1417.3 USD InStock
10048-824 CA10048-824
EarlyTox Cardiotoxicity Kit, Molecular Devices
Assays Cellular Assays
For researchers looking to identify cardiotoxic compounds, the EarlyTox™ Cardiotoxicity Kit allows you to screen more compounds earlier in drug discovery
  • Eliminate cardiotoxic compounds
  • Identify potential drug candidates
  • Reduce well-to-well variation
  • Improve data quality
  • Largest signal dynamic range available
Eliminate cardiotoxic compounds and identify potential drug candidates. Measure 384 samples in minutes rather than hours. Minimize non-specific effect of dye on beat characteristics. Reduce well-to-well variation and improve data quality. Largest signal dynamic range available. Benchmark against three reference compounds: isoproterenol, sotalol and propranolol.
Now you can prioritize leads and direct medicinal chemistry efforts sooner, improving productivity and reducing costs associated with extensive safety testing downstream.
Ordering information: Each Kit is sufficient for 10 plates. Three reference compounds are included (Isoproterenol, Sotalol, Propranolol).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty