Would you like to log in again?
Capillary Column End Caps, Thermo Scientific
Supplier: Thermo Scientific
260EC111
10055-046EA 107.3 USD InStock
10055-046
Capillary Column End Caps, Thermo Scientific
Fittings and Connectors
Eliminate column contamination caused by leaving a column unsealed or sealed with a septum
Color-coordinated fittings ensure that the column is reinstalled the same way it came out. Reusable and universally fitting to all GC capillary columns.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty