Would you like to log in again?
Hypersil™ CPS HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
30805-204630 30805-254630 30803-104630 30805-254030 30803-154630 30805-124030 30805-154630
10042-702EA 1096.73 USD InStock
10042-702 10042-688 10042-700 10042-676 10042-696 10042-694 10042-670
Hypersil™ CPS HPLC Columns, Thermo Scientific
Chromatography Columns
Classical Hypersil stationary phases have global recognition as an industry standard in HPLC, providing an effective analytical tool.
Hypersil phases are well established and referenced in many HPLC methods worldwide.
Certifications: Manufactured under ISO 9001-2000 quality guidelines.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty