Would you like to log in again?
Hypersil™ Silica HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
30003-014001 30005-154630 30005-254630 30005-054630 30005-304030 30003-254630 30003-154630 30005-204630 30003-154030 30005-102130 30005-254030 30005-054030 30005-052130 30005-014001 30005-154030 30005-024006 30005-259070
10045-998EA 1166.55 USD InStock
10045-998 10046-044 10046-000 10046-034 10046-012 10046-020 10046-042 10042-308 10046-032 10046-010 10046-050 10046-008 10046-018 10046-038 10046-016 10046-002 10046-046
Hypersil™ Silica HPLC Columns, Thermo Scientific
Chromatography Columns
Classical Hypersil stationary phases have global recognition as an industry standard in HPLC, providing an effective analytical tool.
Hypersil phases are well established and referenced in many HPLC methods worldwide.
Certifications: manufactured under ISO 9001-2000 quality guidelines.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty