Would you like to log in again?
Hypersil™ BDS Phenyl HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
28905-304030 28905-154630 28905-254030 28905-254630 28903-154630
10042-292EA 1000.07 USD InStock
10042-292 10042-268 10042-302 10042-300 10042-298
Hypersil™ BDS Phenyl HPLC Columns, Thermo Scientific
Chromatography Columns
For a robust column with alternative selectivity to C18 or C8, use Thermo Scientific™ Hypersil BDS Phenyl columns.
These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available. For more challenging separations that require improved chromatographic performance, we recommend the use of Hypersil GOLD columns as an alternative.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm Int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm Int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm Int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty