Would you like to log in again?
Hypersil™ Green PAH Columns, Thermo Scientific
Supplier: Thermo Scientific
31105-014001 31103-104030 31105-154630 31103-152130 31103-053030 31103-102130 31103-104630 31103-014001 31105-104630 31103-154630 31103-153030 31105-253030 31103-012101 31103-103030 31103-013001 31105-254630
10046-144EA 1246.86 USD InStock
10046-144 10046-122 10046-120 10046-150 10046-118 10046-108 10046-116 10046-138 10046-128 10046-106 10046-126 10046-148 10046-104 10046-134 10046-112 10046-124
Hypersil™ Green PAH Columns, Thermo Scientific
Chromatography Columns
For the rapid analysis of polyaromatic hydrocarbons (PAHs), choose Thermo Scientific™ Hypersil Green PAH columns.
Featuring specially tailored alkyl-bonded silica with a high carbon loading, these short, fast columns can analyze 16 PAHs in 4 minutes. Optimized for EPA Method 610: Polynuclear Aromatic Hydrocarbons, these columns provide excellent performance.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty