Would you like to log in again?
Hypersil GOLD™ SAX HPLC Columns, Thermo Scientific
Supplier: Thermo Scientific
26305-154630 26303-154630 26305-014001 26305-254630 26302-102130 26305-104630 26302-152130 26302-052130 26305-013001 26303-052130
10041-880EA 1219.13 USD InStock
10041-880 10041-852 10041-886 10041-854 10041-888 10041-866 10041-900 10041-912 10041-848 10041-904
Hypersil GOLD™ SAX HPLC Columns, Thermo Scientific
Chromatography Columns
Analyze smaller organic molecules, including nucleotides and organic acids, quickly and efficiently with Thermo Scientific™ Hypersil GOLD™ SAX HPLC columns.
These quaternary amine strong anion-exchange columns are highly stable for aqueous and low pH mobile phases. Based on high purity silica bonded with an ion-exchange ligand, Hypersil GOLD™ SAX HPLC columns provide outstanding peak shape and sensitivity.
Ordering information: Guard cartridges require use of corresponding Thermo Scientific UNIGUARD holders. Cartridges with a 1 mm int.Ø require the 1 mm UNIGUARD holder (10046-746), 2.1/3 mm int.Ø require the 2/3 mm UNIGUARD holder (10046-748) and 4/4.6 mm int.Ø require the 4/4.6 mm UNIGUARD holder (10046-744).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty