Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Water nuclease-free, DEPC treated

Supplier: IBI Scientific
Synonyms: Dihydrogen oxide

IB42201 IB42240 IB42200 IB42220 IB42221 IB42211 IB42230 IB42210 IB42222 IB42212 IB42202 IB42231
10065-960CS 1023.7 USD InStock
10065-960 10065-972 10065-970 10065-964 10065-976 10065-962 10065-974 10065-968 10065-958 10065-966 10065-978 10065-956
Water nuclease-free, DEPC treated
Water
IBI DEPC-Treated, Nuclease-Free Water is ideally suited for all applications in molecular biology lab including PCR, RT-PCR, restriction enzyme assays, modifying enzyme assays, transfection, cloning, transformation, and specifically for all RNA assays.

  • DEPC treated to be RNAse free water
  • 0.1 µm sterile-filtered into certified-clean polycarbonate bottles
  • Steam-sterilized (autoclaved twice)

IBI DEPC-Treated Water is manufactured under stringent conditions. The purification process for this product includes continuous deionization, reverse osmosis, UV-treatment, 0.1 µm filtration, followed by steam sterilization in an autoclave. The product is re-autoclaved to ensure the breakdown of DEPC and release of carbon dioxide that may cause the pH to shift.


Formula: H₂O
MW: 18.02 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
Order Now
Specification Test Results
Nuclease Activity (DNase & RNase on 2% Gel) None Observed
Sterility (Biological Activity) None Observed
pH (10ml sample after autoclave) 5.0 - 7.5
Appearance (Visual Inspection) Clear

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty