Would you like to log in again?
PermaYellow/HRP, Diagnostic BioSystems
Supplier: Diagnostic BioSystems
K060 K060 K060-110
10064-852EA 643.38 USD InStock
10064-852 10064-854 CA10064-854
PermaYellow/HRP, Diagnostic BioSystems
Stains Electrophoresis Stains
PermaYellow/HRP is a substrate-chromogen system designed to be used for either IHC or ISH when utilizing horseradish peroxidase
PermaYellow/HRP produces a distinct bright yellow that can easily be distinguished from other stains.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty