Would you like to log in again?
Cole-Parmer® RS-250 Series Reaction Stations, Antylia Scientific
Supplier: Antylia Scientific
99968-04 99968-13 36630-12
75994-940EA 7070.79 USD InStock
75994-940 75994-942 75994-938
Cole-Parmer® RS-250 Series Reaction Stations, Antylia Scientific
Stirrers Magnetic Hotplate Stirrers
Run multiple reactions in a single station, saving time and money.
  • Excellent temperature uniformity across the block
  • Soft-start feature minimizes splashing
Run these reaction stations at the same temperature and stirring rates at several positions simultaneously. They are great for a wide range of applications from simple synthesis to process optimization. High-performance magnetic stirrers ensure maximum coupling between the stir bar in the sample and motor.
Easily set and view temperature and stir rate on the digital display. Use the RS232 or RS485 connections to interface with a robotic workstation or a computer. The well-insulated stations stay cool to the touch for safe handling. PTFE coating on vessel block provides protection from chemical spills.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty