Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
GeneMate ME Agarose
Supplier: GeneMate
D1-ME-500G D1-ME-125G D1-ME-25G
490001-584EA 1200.7 USD InStock
490001-584 490001-582 490001-580
GeneMate ME Agarose
Electrophoresis Gels Agarose and Acrylamide
GeneMate ME Agarose has an intermediate EEO value (higher than GeneMate LE), making it ideal for nucleic acid electrophoresis and protein electrophoresis (serum protein and immunoelectrophoresis.) Sulfate: 0.2%.
  • Nucleic Acid Electrophoresis
  • Serum Protein Electrophoresis
  • Immunoelectrophoresis (IEP)
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty