Would you like to log in again?
GeneMate HGT Agarose
Supplier: GeneMate
D2-LE-25G
490016-268EA 147.18 USD InStock
490016-268
GeneMate HGT Agarose
Electrophoresis Gels Agarose and Acrylamide
GeneMate HGT Agarose has a higher gelling temperature than GeneMate ME and GeneMate LE series agaroses, which gives higher thermal stability to the gels.
low. Sulfate, 0.2%
  • Preparation of Agarose Beads
  • Protein Electrophoresis, Immunoelectrophoresis and Couterelectrophoresis
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty