Would you like to log in again?
Axygen® DNA Markers, Corning
Supplier: Corning
Axygen®
M-DNA-100BP M-DNA-1KB
95043-578EA 89.25 USD InStock
95043-578 95043-576
Axygen® DNA Markers, Corning
Electrophoresis Reagents Molecular Weight Markers
Three DNA markers, Low Range, High Range, and Broad Range that allow fast sizing of a wide range of DNA fragments 125 bp to 10 kb
Each vial has 600 µl of Markers sufficient for up to 125 lanes. DNA Concentration is 1 µg/10 µl. Loading Recommendation, Marker is supplied ready-for-use. Just load 5-10 µl per lane.
Each lot is tested in electrophoresis. The DNA concentration is determined using a spectrophotometer and teh absence of nucleases is confirmed by appropriate tests. Temperature stability is confirmed by burn-in tests where the DNA marker is subject to storage at 65°C and 95% humidity for 2 weeks.
Stable under ambient conditions for as long as 2 years. Precise and consistent when run under conventional electrophoresis conditions. Sharp bands for precise software detection and molecular weight determination. Robust-can be used for fast 10-15 minute electrophoresis runs. Bright reference bands for eas detection and quantification. Ready-To-Use, pre-loaded with Bromophenol Blue dye. Two year shelf life with ambient conditions.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty