Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Whatman™ Puradisc Syringe Filters, RC, Whatman products (Cytiva)

Supplier: Whatman products (Cytiva)
Whatman™ Puradisc™
6757-2502 6757-1304 6756-1304 6758-2502 10462960 6757-1302 6758-2504 6757-1302 6756-1302 6758-2504 6758-1304 6757-1304 6756-1302 10462940 10462940 6758-1304 6756-2504 6758-1302 6756-1304 10462945 6757-2502 10462960 10462950 10462950 6758-2502 10462945 6756-2502 6758-1302 6757-2504 6757-2504 6756-2504
76538-820PK 2870.9 USD InStock
CA11013-158 CA76532-356 CA76538-952 CA11013-152 CA76538-950 76538-820 CA76474-042 CA76474-040 10035-114 76474-038 10035-118 10035-116 76532-354 CA11013-128 CA76538-948 76538-818 CA76538-946 76538-816 76532-358 CA76538-944 76532-356 76538-814 76538-812 CA76538-942 76538-810 76474-042 76474-040 CA11012-958 10035-120 CA76474-038 CA76532-358
Whatman™ Puradisc Syringe Filters, RC, Whatman products (Cytiva)
Filters Syringe Filters
Whatman puradisc syringe filter with RC membrane is an excellent choice for any application requiring a chemically resistant, hydrophilic, low protein binding membrane. This filter can be used for both aqueous and aggressive organic solvents.

  • Regenerated cellulose membrane exhibits low extractables and protein binding
  • Convenient syringe format streamlines filtration workflow and saves time
  • Rugged heat-welded construction is autoclavable
  • Choice of 13 or 25 mm syringe filter diameter; choice of 0.2 or 0.45 μm pore size
  • Inlet, female Luer lock (FLL); outlet: male slip Luer (ML)
  • Integrity testable BP or WBT (‘in-situ’)
  • Sterilization: Autoclave at 121 °C (131 °C max.)

Whatman Puradisc RC Syringe Filters provide rapid, routine syringe filtration of solvents, chemicals, aqueous, and non-aqueous solutions and samples up to 100 mL. They consist of a regenerated cellulose (RC) membrane within a sonicly-welded polypropylene (PP) housing. These rugged, hydrophilic syringe filters can stand up to aggressive organic solvents, while providing a cost-effective solution for rapid filtration in high-throughput labs.

Whatman Puradisc line is a comprehensive syringe filter portfolio used in a variety of applications, including capillary electrophoresis, biological preparations, protein filtration, and UV spectroscopy sample preparation. Combining premium quality with cost efficiency, Whatman Puradisc syringe filters are well suited for rapid, routine syringe filtration of samples up to 100 ml.

Puradisc filters are available in a range of diameters and pore sizes, and eleven different material options to support diverse chemical solvents, compounds and sample types. Additional options include sterile, medical grade blister pack and tube tip format for accurate dispensing into a micro vial.

Whatman also offers high-performance GD/X syringe filters with a multi-layer glass fiber prefilter for difficult-to-filter samples.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty