Would you like to log in again?
Versa-Top Universal Fit Tips
Supplier: Scientific Specialties
4110-00-120IS
490015-170BG 40.45 USD InStock
490015-170
Versa-Top Universal Fit Tips
Pipette Tips
These tips are available in a standard lift top rack, or a dual-position hinged-lid rack that will either rotate back, or lift off for those who prefer to remove the lid.
All beveled tips offer an excellent fit on Labpette, Pipetman, Finnpipette, Oxford Benchmate, and Sealpette pipettors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty