Would you like to log in again?
Fast-Ion Midi Plasmdi Kits, IBI Scientific
Supplier: IBI Scientific
IB47111
95039-658EA 368.97 USD InStock
95039-658
Fast-Ion Midi Plasmdi Kits, IBI Scientific
IBI’s Midi Plasmid Kits use pre-packed ion-exchange resin columns to purify plasmid or cosmid DNA from bacterial cultures
The plasmid DNA is suitable for transfection, sequencing reactions, PCR, microinjection, restriction digestion, and in-vitro transcription.
In this process, the modified alkaline lysis method and RNase treatment are used for creating cleared cell lysate with minimal genomic DNA/RNA contaminants. Using a gravity-flow procedure, the plasmid DNA in crude lysate binds to the column. The contaminants are washed away with a wash buffer. The purified plasmid DNA is eluted by a high salt buffer and precipitated with isopropanol. The entire procedure can be completed in less than 2 hours.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty