Would you like to log in again?
Thick Chromatography Paper, Grade 243, Ahlstrom
Supplier: Ahlstrom
2438-4657
47749-850PK 1072.3 USD InStock
47749-850
Thick Chromatography Paper, Grade 243, Ahlstrom
Blotting Paper
Ahlstrom Grade 243 is one of the thicker chromatography papers available from Ahlstrom
It offers moderately high flow and absorption and is recommended for chromatography with heavy solute loading.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty