Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 3B9.E6]

Supplier: Rockland Immunochemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

200-301-F56
RL200-301-F56EA 532 USD InStock
RL200-301-F56
Anti-DNAJB1 Mouse Monoclonal Antibody [clone: 3B9.E6]
Antibodies
Anti-Hsp40 Antibody is recommended for use in WB, IP, Immunochemistryand ELISA. Expect a band approximately ~40kDa Protein corresponding to the molecular mass of Hsp40 on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1:2000;

Type: Primary
Antigen: DNAJB1
Clonality: Monoclonal
Clone: 3B9.E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse, Rat, Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty