Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL21 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-B53S 200-401-B53
RL200-401-B53EA 518 USD InStock
RL200-401-B53 RL200-401-B53S
Anti-IL21 Rabbit Polyclonal Antibody
Antibodies
IL-21 is expressed in activated CD4-positive T-cells but not in CD8-positive T-cells, B-cells, or monocytes, and in HL-60 and THP-1 cell lines. This Protein-A purified antibody has been tested for use in ELISA and western blotting.

Recommended Dilutions: ELISA: 1:6000; Western Blot: 1 µg/ml;

Type: Primary
Antigen: IL21
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig, Horse, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty