Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSF2 Rat Monoclonal Antibody [clone: 300]

Supplier: Rockland Immunochemicals
200-501-F51
RL200-501-F51EA 532 USD InStock
RL200-501-F51
Anti-HSF2 Rat Monoclonal Antibody [clone: 300]
Antibodies
Anti-HSF2 Antibody is suitable for WB. Expect a band approximately ~69kDa Protein in unstressed cells corresponding to the molecular mass HSF2 on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 4µg/ml

Type: Primary
Antigen: HSF2
Clonality: Monoclonal
Clone: 300
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Guinea Pig, Hamster, Monkey, Rabbit Dog, Bovine, Sheep, Pig

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty