Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP2 Chicken Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-901-D68
RL200-901-D68EA 534 USD InStock
RL200-901-D68
Anti-MAP2 Chicken Polyclonal Antibody
Antibodies
Anti-Microtubule Associated Protein 2 antibody is suitable for use in Western Blotting and IF. Specific conditions should be optimized by the end user. Expect a band of approximately 280 kDa in size corresponding to MAP2 Proteins in Western blots.

Recommended Dilutions: Immunofluorescence: 1:2500; Western Blot: 1:20000;

Type: Primary
Antigen: MAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty