Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IL1B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
209-401-B73
RL209-401-B73EA 518 USD InStock
RL209-401-B73
Anti-IL1B Rabbit Polyclonal Antibody
Antibodies
Anti-Human IL-1-beta has been tested for use in neutralizations, ELISA, radioimmunoassays, flow cytometry.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: IL1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Primate, Dog

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty