Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-EBI3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Rockland Immunochemicals
210-403-B66
RL210-403-B66EA 498 USD InStock
RL210-403-B66
Anti-EBI3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
IL-27 is expressed in activated antigen presenting cells including monocytes, endothelial cells, and dendritic cells, for example mouse CD4 splenocytes. This purified antibody has been tested for use in western blotting.

Recommended Dilutions: ELISA: 1:10000-1:50000; Western Blot: 1:1000-1:5000; Immunochemistry: 1:500-1:2500
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: EBI3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty