Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG1 gamma Goat Polyclonal Antibody (ATTO 647N)

Supplier: Rockland Immunochemicals
610-156-040
RL610-156-040EA 405.61 USD InStock
RL610-156-040
Anti-IgG1 gamma Goat Polyclonal Antibody (ATTO 647N)
Antibodies
The emission spectra for this ATTO conjugate matches the principle output wavelengths of most common fluorescence instrumentation.

Recommended Dilutions: FLISA: >1:20000; Immunofluorescence: >1:5000; Flow Cytometry: 1:500 - 1:2500; Western Blot: >1:10000;
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG1 gamma
Clonality: Polyclonal
Clone:
Conjugation: ATTO 647N
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty