Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Rockland Immunochemicals
611-702-003
RL611-702-003EA 177 USD InStock
RL611-702-003
Anti-IgG Donkey Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Recommended Dilutions: FLISA: 1:100000; Immunofluorescence: 1:1000 - 1:5000; Western Blot: 1:2000 - 1:20000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: F(c)
Host: Donkey
Isotype:
Reactivity: Rabbit

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty