Would you like to log in again?
Caps for Nunclon™ Δ Flasks, Thermo Scientific
Supplier: Thermo Scientific
144458 144652
73520-538CS 112.47 USD InStock
73520-538 73520-520
Caps for Nunclon™ Δ Flasks, Thermo Scientific
Closures Bottle Closures
Replacement caps for Nunclon Delta flasks.
  • Filter caps allow constant airflow and minimize chance of contamination
  • Vent/close caps feature a "Y" mark that allows visual verification of vent position
For use with 80 cm2 flasks.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty