Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Anti-PAX8 Mouse Monoclonal Antibody [clone: BC12]

Supplier: Diagnostic BioSystems

PDM180
10065-004EA 445.52 USD InStock
10065-004
Anti-PAX8 Mouse Monoclonal Antibody [clone: BC12]
Antibodies
PAX 8 is expressed in a high percentage of renal cell carcinomas and ovarian cancers. This mouse monoclonal PAX 8 antibody [BC12] has been designed to target restricted epitopes, and exhibits higher specificity and provides sharper staining than the PAX 8 rabbit polyclonal antibody. Unlike the polyclonal PAX 8, this mouse monoclonal antibody does not stain B-cells, and does not recognize epitopes of pancreatic origin and neuroendocrine cells in stomach and colon; thus providing superior specificity. The expression of the mouse monoclonal PAX 8 targets antigens found in normal kidney, thyroid and cervix, but not normal ovary. PAX 8 stains nuclei exclusively and performs well in formalin-fixed paraffin-embedded tissues.

Type:
Antigen: PAX8
Clonality: Monoclonal
Clone: BC12
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: 0

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty