Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Renal Cell Carcinoma Mouse Monoclonal Antibody [clone: PN-15]

Supplier: Diagnostic BioSystems

MOB465 MOB465-05 PDM169
10064-990EA 302.4 USD InStock
10064-990 10064-946 10064-948
Anti-Renal Cell Carcinoma Mouse Monoclonal Antibody [clone: PN-15]
Antibodies
This antibody recognizes a 200 kD glycoprotein (gp200) localized in the brush border of the proximal renal tebules and the luminal surface of Bowman’s capsule. gp200 was expressed by 93% of primary and 84% of metastatic renal carcinomas. Gp200 glycoprotein is also localized in the breast lobules and ducts, parathyroid glands and epididymis.

Type:
Antigen: Renal Cell Carcinoma
Clonality: Monoclonal
Clone: PN-15
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty