Would you like to log in again?
Whatman™ Grade 802 Qualitative Filter Papers, Whatman products (Cytiva)
Supplier: Whatman products (Cytiva)
Whatman™
5802-320 5802-6698 5802-385 5802-150 5802-240 5802-385 5802-320 5802-185 5802-6698 5802-185 5802-125 5802-125 5802-150 5802-240
70240-110PK 59.43 USD InStock
70240-110 CA11013-874 70240-112 CA70240-110 70240-114 CA70240-112 CA28467-606 CA70240-114 CA28467-607 10035-482 28467-607 28467-606 CA28467-611 28467-611
Whatman™ Grade 802 Qualitative Filter Papers, Whatman products (Cytiva)
Filters Filter Paper Qualitative Filter Paper
A prepleated filter for use with a conical filter funnel, offers fast filtration and high loading capacity for analysis involving coarse particles or gelatinous precipitates.
  • Highest retention among standard filters
  • Slow flow rate
  • Fine-particle retention
  • For qualitative analysis
The filter is wet-strengthened and for normal qualitative application it will not introduce any significant impurities into the filtrate. However, it is not recommended for Kjeldahl nitrogen analysis.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty