Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HSPA4 Rabbit Polyclonal Antibody

Supplier: Prosci
26-101
10103-142EA 745.96 USD InStock
10103-142
Anti-HSPA4 Rabbit Polyclonal Antibody
Antibodies
HSPA4 (Hsp70) belongs to the heat shock protein 70 family. It was isolated as a putative Rictor interacting protein and interaction with membranes acts as a platform for its release into the extracellular environment during its recovery from stress. Hsp70 gene expression in Rheumatoid Arthitis-affected synovial tissue is followed by Hsp70 cell surface expression on fibroblast-like synovial cells growing from RA synovial tissue. Serum Hsp70 levels were increased in Systemic sclerosis patients, and associated with pulmonary fibrosis, skin sclerosis, renal vascular damage, oxidative stress, and inflammation.

HSPA4 antibody can be used for detection of HSPA4 by ELISA at 1:62500. HSPA4 antibody can be used for detection of HSPA4 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: HSPA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Dog

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty