Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PFDN6 Rabbit Polyclonal Antibody

Supplier: ProSci
26-643
10103-928EA 745.96 USD InStock
10103-928
Anti-PFDN6 Rabbit Polyclonal Antibody
Antibodies
PFDN6 binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. PFDN6 binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.

PFDN6 antibody can be used for detection of PFDN6 by ELISA at 1:1562500. PFDN6 antibody can be used for detection of PFDN6 by western blot at 1 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PFDN6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty