Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ZRSR2 Rabbit Polyclonal Antibody

Supplier: ProSci
29-347
10108-078EA 680.93 USD InStock
10108-078
Anti-ZRSR2 Rabbit Polyclonal Antibody
Antibodies
ZRSR2 is an essential splicing factor. The protein associates with the U2 auxiliary factor heterodimer, which is required for the recognition of a functional 3' splice site in pre-mRNA splicing, and may play a role in network interactions during spliceosome assembly.This gene encodes U2AF small subunit related protein 2 which is an essential splicing factor. This protein is associated with the U2AF heterodimer composed of a large and a small subunit. It specifically interacts with the large subunit through an U2AF small subunit homologous region, and with SR proteins through its RS domain. This protein and U2AF small subunit are not functionally redundant.

ZRSR2 antibody can be used for detection of ZRSR2 by ELISA at 1:62500. ZRSR2 antibody can be used for detection of ZRSR2 by western blot at 2.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ZRSR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty