Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CHI3L1 Rabbit Polyclonal Antibody

Supplier: ProSci
30-782
10110-698EA 745.96 USD InStock
10110-698
Anti-CHI3L1 Rabbit Polyclonal Antibody
Antibodies
CHI3L1 may play an important role in the capacity of cells to respond to and cope with changes in their environment. CHI3L1 may play a role in tissue remodeling and defense against pathogens. It belongs to the glycosyl hydrolase 18 family. CHI3L1 may play an important role in the capacity of cells to respond to and cope with changes in their environment.

CHI3L1 antibody can be used for detection of CHI3L1 by ELISA at 1:312500. CHI3L1 antibody can be used for detection of CHI3L1 by western blot at 0.5 ?g/mL, and HRP conjugated secondary antibody should be diluted 1:50,000 - 100,000.

Antibody is purified by peptide affinity chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 ?L of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CHI3L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty