Would you like to log in again?
Human Recombinant IFN beta (from CHO cells)
Supplier: ProSci
40-278
10072-864EA 302.43 USD InStock
10072-864
Human Recombinant IFN beta (from CHO cells)
Proteins and Peptides
Proteins of this family play an important role in inducing non-specific resistance against a broad range of viral infections. They also affect cell proliferation and modulate immune responses. Produced by peripheral blood leukocytes and lymphoblastoid cells, IFNα is an acid stable molecule that signals through IFNα/βR, which is also used by IFNβ. Both IFNs have similar anti-viral activity and regulate expression of MHC class I antigens. IFNα contains four highly conserved cysteine residues which form two disulfide bonds, one of which is necessary for biological activity. Recombinant human IFNβ is an 20.0 kDa protein containing 166 amino acid residues. Due to glycosylation, IFNβ has an approximate MW of 22.3 kDa based on SDS-PAGE gel and Mass Spectrometry.
Store in working aliquots at -20°C. Avoid freeze/thaw cycles.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty