Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vikan® Fiberglass Handles, 59", Remco Products

Supplier: Vikan
29384 29389 29386 29383 29381 29388 29382 29385 29387
76007-258EA 39.36 USD InStock
76007-258 76007-260 76007-264 76007-252 76007-262 76007-250 76007-256 76007-266 76007-254
Vikan® Fiberglass Handles, 59", Remco Products
Brooms
This standard broom handle can be used with all brooms, squeegees and scrapers.

Aluminum and polypropylene materials

The handle with the attached item should reach to the chin of the user to prevent occupational back injuries.

Width: 1.25"

Certifications: FDA compliant raw material.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty