Would you like to log in again?
Vikan® Hand Scrub Brushes, Stiff, Remco Products
Supplier: Remco
Vikan®
38926 38925 38923 38924
76007-392EA 38.79 USD InStock
76007-392 76007-390 76007-394 76007-388
Vikan® Hand Scrub Brushes, Stiff, Remco Products
Brushes
This large hand scrub brush with flared bristles enables you to clean tables, chopping boards, buckets, large bowls and other equipment where it is necessary to reach into corners.
  • Block material is polypropylene; bristle material is polyester
  • Dishwasher safe
  • Autoclavable up to 250°F
Width: 4.75''
Height: 2.4''
Certifications: FDA compliant raw material.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty