Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Power Supply, AC/DC 30 V

Supplier: EISCO SCIENTIFIC LLC
PWS30V
470138-950EA 344.35 USD InStock
470138-950
Power Supply, AC/DC 30 V
Power Supplies
This regulated DC power supply can be used for any general purpose requirement.

  • Continuously adjustable DC output from 0 to 30 V DC at 2 A
  • Standard AC output 1 to 6 V, 1 V increments
  • Overload and short circuit protected

This highly versatile supply comes with digital displays for both current and voltage. Housed in high quality ABS case, each of the outputs are protected from overload and short circuit. This unit is perfect for driving motors, digital and analog circuits, light sources and many more. The outputs include 1-6 V AC, 2 Amp max in steps of 1 V, and 0-30V DC with 2 Amp max, which is adjustable. Input to the unit is switchable 120 V/60 Hz and 240 V, 50/60 Hz.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty