Would you like to log in again?
GOLD BRAND ASTM/API High Precision Combined Form Hydrometer, Thermco
Supplier: Thermco
ACC058HLS ACC052HLS ACC059HLS ACC060HLS ACC055HLS ACC054HLS ACC057HLS ACC051HLS ACC053HLS ACC056HLS
10053-120EA 112.99 USD InStock
10053-120 10053-122 10053-110 10053-124 10053-112 10053-114 10053-106 10053-116 10053-108 10053-118
GOLD BRAND ASTM/API High Precision Combined Form Hydrometer, Thermco
Hydrometers
High precison ASTM-like combined form hydrometers made to exceed ASTM and API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 2.0 degrees farenheit. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 0 to 150 degrees farenheit and is mounted in the body of the hydrometer. Length of the hydrometer is 380mm.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty