Would you like to log in again?
GOLD BRAND ASTM/API High Precision Combined Form +30 / 220 °F Hydrometer, Thermco
Supplier: Thermco
ACC071HS ACC074HLS ACC073HS ACC072HLS
10053-130EA 130.14 USD InStock
10053-130 10053-132 10053-126 10053-128
GOLD BRAND ASTM/API High Precision Combined Form +30 / 220 °F Hydrometer, Thermco
Hydrometers
High precison ASTM-like combined form hydrometers made to exceed ASTM and API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 2.0 degrees farenheit. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 30 to 220 degrees farenheit and is mounted in the hydrometer stem. Length of hydrometer is 380 mm.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty